Skip to Content
Merck
All Photos(1)

Key Documents

1370600

USP

Lovastatin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Mevinolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lovastatin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O1[C@@H](C[C@H](CC1=O)O)CC[C@@H]2[C@H]3[C@H](C[C@H](C=C3C=C[C@@H]2C)C)OC(=O)[C@H](CC)C

InChI

1S/C24H36O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,10,14-16,18-21,23,25H,5,8-9,11-13H2,1-4H3/t14-,15-,16-,18+,19+,20-,21-,23-/m0/s1

InChI key

PCZOHLXUXFIOCF-BXMDZJJMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lovastatin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Lovastatin Tablets
  • Simvastatin
  • Simvastatin Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrico Peiretti et al.
Experimental eye research, 124, 11-16 (2014-05-06)
Starting from previous studies showing that patients with cognitive deficit present neutral lipids (NLs) accumulation in cytoplasm of their peripheral blood mononuclear cells (PBMCs) and considering that there is epidemiological evidence linking age-related macular degeneration (AMD) to cognitive deficit, the
Dae-Hyoung Yoo et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1508-1513 (2014-06-21)
Orally administered drugs may be metabolized by intestinal microbial enzymes before absorption into the blood. Accordingly, coadministration of drugs affecting the metabolic activities of gut microbes (e.g., antibiotics) may lead to drug-drug interactions (DDI). In this study, gut microbiota-mediated DDI
Takashi Ohkura et al.
Journal of virology, 88(17), 10039-10055 (2014-06-27)
In polarized epithelial cells, influenza A virus hemagglutinin (HA) and neuraminidase (NA) are intrinsically associated with lipid rafts and target the apical plasma membrane for viral assembly and budding. Previous studies have indicated that the transmembrane domain (TMD) and cytoplasmic
Xin Yang et al.
The Journal of general virology, 95(Pt 6), 1330-1337 (2014-03-20)
We examined the role of HMG-CoA reductase (HMGCR) during porcine circovirus 2 (PCV2) infection. The results demonstrated that levels of endogenous HMGCR were not significantly different in PCV2-infected cells and mock-infected cells. However, the level of phosphorylated HMGCR, an inactivated
D S Alejandro et al.
Journal of the American Society of Nephrology : JASN, 5(2), 153-160 (1994-08-01)
Acute renal failure can occur in cardiac transplant patients for a variety of reasons. A case of a patient who developed acute renal failure secondarily to drug-induced rhabdomyolysis is reported. The literature regarding acute renal failure and lovastatin and other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service