Skip to Content
Merck
All Photos(1)

Documents

1357001

USP

L-Leucine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

leucine

manufacturer/tradename

USP

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

ʟ-Leucine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Srinivas R B Behara et al.
Journal of pharmaceutical sciences, 103(2), 465-477 (2013-12-07)
High-efficiency dry powder inhalers (DPIs) were developed and tested for use with carrier-free formulations across a range of different inhalation flow rates. Performance of a previously reported DPI was compared with two new designs in terms of emitted dose (ED)
Xiang Li et al.
International journal of cardiology, 183, 198-208 (2015-02-11)
Evidence from human and animal studies has demonstrated elevated levels of the cysteine protease cathepsin S (CatS) in hypoxic atherosclerotic lesions. We hypothesized that silencing of CatS gene would suppress ischemia-induced angiogenic action. Left femoral artery ligation-induced ischemia in mice
Xiaodi Fan et al.
Journal of translational medicine, 13, 92-92 (2015-04-18)
Inflammation induced by oxidized low-density lipoprotein (ox-LDL) plays an important role in the pathogenesis of atherosclerosis. Recently, roles of autophagy against inflammation in the process of atherosclerosis have drawn increasing attention. Here, we tested the possible molecular mechanisms by which
Yanhui Xu et al.
PloS one, 10(2), e0117081-e0117081 (2015-02-07)
Enhanced activity of interleukin 17 (IL-17) producing T helper 17 (Th17) cells plays an important role in autoimmune and inflammatory diseases. Significant loss of body weight and appetite is associated with chronic inflammation and immune activation, suggesting the cross talk
Abderrazak Kitsy et al.
PloS one, 9(7), e102615-e102615 (2014-07-23)
The essential branched-chain amino acids (BCAA), leucine, valine and isoleucine, are traditionally associated with skeletal muscle growth and maintenance, energy production, and generation of neurotransmitter and gluconeogenic precursors. Recent evidence from human and animal model studies has established an additional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service