Skip to Content
Merck
All Photos(1)

Key Documents

1343550

USP

Iodixanol Related Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Iopentol, 5-[Acetyl(2-hydroxy-3-methylpropyl)amino]-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28I3N3O9
CAS Number:
Molecular Weight:
835.16
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iodixanol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C20H28I3N3O9/c1-9(29)26(5-12(32)8-35-2)18-16(22)13(19(33)24-3-10(30)6-27)15(21)14(17(18)23)20(34)25-4-11(31)7-28/h10-12,27-28,30-32H,3-8H2,1-2H3,(H,24,33)(H,25,34)

InChI key

IUNJANQVIJDFTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iodixanol Related Compound D USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Iodixanol Injection
  • Iodixanol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Jung et al.
Journal of computer assisted tomography, 24(5), 783-787 (2000-10-25)
To evaluate the diagnostic value of MR imaging in abdominal lymphoma staging, in comparison with that of the established spiral CT, within the context of a prospective study. 50 patients with non-Hodgkin (n = 27) and Hodgkin lymphoma (n =
H U Kauczor et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 174(3), 357-362 (2002-03-09)
Determination of the intrahepatic distribution volume of two contrast media (CM) by CT-guided application in an ex-vivo and an in-vivo model (pig liver). In pig livers ex-vivo and in-vivo, 131 CT-guided injections of two different CM (Imagopaque(R), Visipaque(R)) were performed
Markus Selzner et al.
Annals of surgery, 240(6), 1027-1034 (2004-12-01)
To compare the diagnostic value of contrast-enhanced CT (ceCT) and 2-[18-F]-fluoro-2-deoxyglucose-PET/CT in patients with metastatic colorectal cancer to the liver. Despite preoperative evaluation with ceCT, the tumor load in patients with metastatic colorectal cancer to the liver is often underestimated.
S G Spitzer et al.
Acta radiologica (Stockholm, Sweden : 1987), 43(6), 617-622 (2002-12-18)
To investigate the influence of two non-ionic radiographic contrast media with different osmolality on thrombocytic function and the plasmatic coagulation system. The study was carried out as a randomised, prospective, comparative study with two contrast media in a heart catheter
Nikolaos L Kelekis et al.
Clinical imaging, 28(2), 124-127 (2004-03-31)
Confluent hepatic fibrosis (CHF) is usually a feature of advanced cirrhosis, while occurrence in early stage compensated cirrhosis is uncommon. We report the imaging findings of masslike CHF in a patient with previously unsuspected compensated alcoholic cirrhosis. The combination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service