Skip to Content
Merck
All Photos(1)

Key Documents

1304016

USP

Heparin sodium for assays

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Heparin sodium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

heparin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[S](=O)(=O)(NC1C(OC(C(C1O[S](=O)(=O)O)OC4OC(C(C(C4O[S](=O)(=O)O)O)O)C(=O)O)CO)OC2C(OC(C(C2O)O[S](=O)(=O)O)OC3C(OC(C(C3O)NC(=O)C)O)CO[S](=O)(=O)O)C(=O)O)O

InChI

1S/C26H42N2O37S5.Na/c1-4(30)27-7-9(31)13(6(56-23(7)39)3-55-67(43,44)45)58-26-19(65-70(52,53)54)12(34)16(20(62-26)22(37)38)60-24-8(28-66(40,41)42)15(63-68(46,47)48)14(5(2-29)57-24)59-25-18(64-69(49,50)51)11(33)10(32)17(61-25)21(35)36;/h5-20,23-26,28-29,31-

InChI key

JRTRSJGZMRQDHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heparin Sodium is the sodium salt of sulfated glycosaminoglycans present in mammalian tissues, which has properties that can prevent blood clotting. It is usually obtained from the intestinal mucosa or other suitable tissues of domestic mammals used for food by humans. Heparin sodium is commonly administered for the prevention of deep venous thrombosis in both medical and surgical patients.
Heparin sodium for assays USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Heparin sodium for assays USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Anticoagulant Heparin Solution
  • Heparin Sodium
  • Antithrombin III Human
  • Protamine Sulfate

Other Notes

This product is part of the USP Biologics program.

Disclaimer

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heparin-induced hyperkalemia
Edes T E and Sunderrajan E V
Archives of Internal Medicine (1985)
Protamine Sulfate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(1), 3757-3757 (2017)
?208? Anti-factor Xa and anti-factor IIa assays for unfractionated and low molecular weight heparins
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(2), 6611-6611 (2020)
Anticoagulant Heparin Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 2194-2194 (2019)
Antithrombin III Human
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 355-355 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service