Skip to Content
Merck
All Photos(1)

Key Documents

1181302

USP

Dextrose

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-(+)-Glucose, Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dextrose

manufacturer/tradename

USP

mp

150-152 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextrose USP reference standard for specified quality tests and assay use.

Also used to prepare standard and system suitability solutions for the assay and identification tests according to the given below monographs of United States Pharmacopeia (USP):
  • Dextrose
  • Dextrose Excipient
  • Liquid Glucose
  • Galactose
  • Dextrates

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Galactose
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(5) (2020)
Dextrates
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 47(1), 5742-5742 (2018)
Dextrose Excipient
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(5), 5743-5743 (2020)
Dextrose
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(6), 1321-1321 (2020)
Liquid Glucose
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 49(1), 5788-5788 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service