Skip to Content
Merck
All Photos(1)

Documents

1141024

USP

Clotrimazole related compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(o-Chlorophenyl)diphenylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15ClO
CAS Number:
Molecular Weight:
294.77
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clotrimazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C19H15ClO/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,21H

InChI key

KTVAHLGKTSPDOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clotrimazole-related compound A USP reference standard suitable for use in specified USP compendial tests and assays.

Also used to prepare standard stock, system suitability, and standard solution for assay and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Clotrimazole
  • Clotrimazole Topical Solution
  • Clotrimazole Lotion

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clotrimazole Lotion
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(2), 1119-1119 (2013)
Clotrimazole Topical Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(3), 1122-1122 (2016)
Clotrimazole
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(3), 1118-1118 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service