Skip to Content
Merck
All Photos(1)

Key Documents

1134062

USP

Cimetidine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

SKF-92334

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N6S
CAS Number:
Molecular Weight:
252.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cimetidine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN\C(NC#N)=N\CCSCc1nc[nH]c1C

InChI

1S/C10H16N6S/c1-8-9(16-7-15-8)5-17-4-3-13-10(12-2)14-6-11/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14)

InChI key

AQIXAKUUQRKLND-UHFFFAOYSA-N

Gene Information

human ... HRH2(3274)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cimetidine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Xing Yue et al.
CNS neuroscience & therapeutics, 20(10), 883-892 (2014-07-06)
The upregulation of Nav1.8 in primary afferents plays a critical role in the development and persistence of neuropathic pain. The mechanisms underlying the upregulation are not fully understood. The present study aims to investigate the regulatory effect of histamine on
Catherine H Olson et al.
Journal of biomedical informatics, 51, 60-71 (2014-04-17)
Create an automated algorithm for predicting elderly patients' medication-related risks for readmission and validate it by comparing results with a manual analysis of the same patient population. Outcome and Assessment Information Set (OASIS) and medication data were reused from a
Zhuo Wang et al.
Cancer chemotherapy and pharmacology, 74(2), 239-247 (2014-06-01)
We investigated E-selectin expression in mice and rabbits with vinorelbine-induced phlebitis and the effect of cimetidine. To find the relationship between E-selectin expression and vinorelbine-induced phlebitis. Mouse and rabbit model of vinorelbine-induced phlebitis was established by intravenous infusion of vinorelbine.
Ryan J Schutte et al.
Journal of neurophysiology, 112(4), 903-912 (2014-05-09)
Hundreds of mutations in the SCN1A sodium channel gene confer a wide spectrum of epileptic disorders, requiring efficient model systems to study cellular mechanisms and identify potential therapeutic targets. We recently demonstrated that Drosophila knock-in flies carrying the K1270T SCN1A
Naoki Ito et al.
Journal of pharmaceutical sciences, 103(10), 3342-3348 (2014-09-02)
Drug transfer into milk is a general concern during lactation. So far, breast cancer resistance protein (Bcrp) is the only transporter known to be involved in this process, whereas participation of other transporters remains unclear. We investigated the importance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service