1067760
USP
Betamethasone Dipropionate Related Compound F
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
9,11-Epoxy-16-methyl-3,20-dioxopregna-1,4-diene-17,21-diyl dipropionate
About This Item
Recommended Products
API family
betamethasone
packaging
pkg of 30 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
O1[C@@]32[C@@H]1C[C@]4([C@H]([C@@H]3CCC5=CC(=O)C=C[C@@]52C)C[C@@H]([C@]4(OC(=O)CC)C(=O)COC(=O)CC)C)C
InChI
1S/C28H36O7/c1-6-23(31)33-15-21(30)27(35-24(32)7-2)16(3)12-20-19-9-8-17-13-18(29)10-11-25(17,4)28(19)22(34-28)14-26(20,27)5/h10-11,13,16,19-20,22H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,22-,25-,26-,27-,28+/m0/s1
InChI key
BWSCDDHHJUGBSQ-XHYJHYNESA-N
General description
Application
Analysis Note
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B - STOT RE 2
Target Organs
Liver,Kidney,Endocrine system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service