Skip to Content
Merck
All Photos(1)

Key Documents

1029002

USP

Amitriptyline hydrochloride

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N · HCl
CAS Number:
Molecular Weight:
313.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

amitriptyline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CN(C)CC\C=C1\c2ccccc2CCc3ccccc13

InChI

1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H

InChI key

KFYRPLNVJVHZGT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amitriptyline hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amitriptyline Hydrochloride Tablets
  • Chlordiazepoxide and Amitriptyline Hydrochloride Tablets
  • Cyclobenzaprine Hydrochloride Extended-Release Capsules
  • Perphenazine and Amitriptyline Hydrochloride Tablets

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the norepinephrine and serotonin transporters with Kis of 100 nM and 14.7 nM, respectively; high in vitro affinity for α1-adrenoceptors, serotonin and muscarinic acetylcholine receptors.
Tricyclic antidepressant; norepinephrine and serotonin transporter inhibitor; high in vitro affinity for α1-adrenoceptors, serotonin and muscarinic acetylcholine receptors.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katja Kovacic et al.
The Journal of pediatrics, 164(5), 1104-1109 (2014-03-13)
To compare children with primary, chronic idiopathic nausea to those with secondary nausea associated with functional abdominal pain. Retrospective chart review of 45 children with a primary complaint of chronic nausea several times per week. Comparisons were made to prospectively
Jennifer S Gewandter et al.
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer, 22(7), 1807-1814 (2014-02-18)
Chemotherapy-induced peripheral neuropathy (CIPN) occurs in as high as 70% of patients receiving certain types of chemotherapy agents. The FDA has yet to approve a therapy for CIPN. The aim of this multicenter, phase III, randomized, double-blind, placebo-controlled trial was
Mariola Herbet et al.
Acta poloniae pharmaceutica, 70(5), 913-917 (2013-10-24)
The aim of the present study was to assess the impact of combined therapy with rosuvastatin (10 mg/kg) and amitriptyline (10 mg/kg) on oxidation-reduction status in the blood of rats. After 2-week application of drugs alone or their combination, the
Grzegorz Jagla et al.
Pharmacological reports : PR, 66(3), 459-465 (2014-06-07)
The therapy of neuropathic pain may include the use of co-analgesics, such as antidepressants, however, their desired analgesic effect is associated with significant side effects. An alternative approach to this is their local administration which has been proposed, but there
Ming Yang et al.
Aquatic toxicology (Amsterdam, Netherlands), 151, 68-76 (2014-01-23)
Amitriptyline is a tricyclic antidepressant used for decades. It is present at low detectable concentrations in the aquatic environment, but relative few studies have focused on its ecotoxicological effects on non-target aquatic animals. The present study conducted an acute toxicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service