Skip to Content
Merck
All Photos(1)

Key Documents

1012553

USP

Albendazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl 5-(propylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O2S
CAS Number:
Molecular Weight:
265.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

albendazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCSc1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

HXHWSAZORRCQMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Albendazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Albendazole Oral Suspension
  • Albendazole Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,Blood,male reproductive organs,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Horton
Parasitology, 121 Suppl, S113-S132 (2001-06-02)
This comprehensive review briefly describes the history and pharmacology of albendazole as an anthelminthic drug and presents detailed summaries of the efficacy and safety of albendazole's use as an anthelminthic in humans. Cure rates and 0% egg reduction rates are
Matthew E Falagas et al.
The American journal of the medical sciences, 334(3), 171-179 (2007-09-18)
Albendazole has been used in various ways in the treatment of cystic echinococcosis (CE). We reviewed the available evidence regarding the role of albendazole for the treatment of patients with CE. The available comparative clinical trials (randomized or not) that
M E Oxberry et al.
Journal of veterinary pharmacology and therapeutics, 23(3), 113-120 (2000-12-08)
Trophozoites of the protozoan parasite Giardia duodenalis were exposed to various albendazole concentrations for 4 h, washed, fixed and incubated with antibodies raised against albendazole and its two major metabolites albendazole sulphoxide and albendazole sulphone. Tubulin antibodies were also used.
John Horton
Fundamental & clinical pharmacology, 17(2), 205-212 (2003-04-02)
Twenty years after the first cases of echinococcal disease were treated with albendazole, it has become an important component in the overall management of both cystic and alveolar echinococcosis. This paper reviews some of the key steps and issues facing
Marlieke E A de Kraker et al.
Tropical medicine & international health : TM & IH, 11(5), 718-728 (2006-04-28)
To determine the efficacies of combinations of ivermectin or diethylcarbamazine and albendazole, which are recommended for use in mass treatment programmes against lymphatic filariasis. Review of published trends in microfilarial (mf) intensities after treatment with these combination therapies. By fitting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service