Skip to Content
Merck
All Photos(1)

Key Documents

1011029

USP

Acitretin Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ethyl all-trans-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoate, Etretinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O3
CAS Number:
Molecular Weight:
354.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acitretin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(C)c1c(c(c(c(c1)C)\C=C\C(=C\C=C\C(=C\C(=O)OCC)\C)\C)C)C

InChI

1S/C23H30O3/c1-8-26-23(24)14-17(3)11-9-10-16(2)12-13-21-18(4)15-22(25-7)20(6)19(21)5/h9-15H,8H2,1-7H3/b11-9+,13-12+,16-10+,17-14+

InChI key

HQMNCQVAMBCHCO-DJRRULDNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acitretin Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Acitretin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoleukomelanoderma possibly caused by etretinate in a patient with psoriasis.
Mariko Seishima et al.
Acta dermato-venereologica, 90(1), 85-86 (2010-01-29)
Hyung-Doo Park et al.
Transfusion, 48(11), 2395-2400 (2008-07-29)
Acitretin and etretinate are potentially teratogenic. Many people taking acitretin for psoriasis have donated blood during the deferral period in Korea. Therefore, many of the blood products from these donors treated with acitretin have been circulated in Korea. A high-performance
Genetically proven Mal de Meleda complicated by Bowen's disease of the sole.
Athanasia Tourlaki et al.
European journal of dermatology : EJD, 21(2), 292-294 (2011-03-09)
Cutaneous gamma/delta T-cell lymphoma treated with retinoid and narrowband ultraviolet B.
H Nakashima et al.
Clinical and experimental dermatology, 34(7), e345-e346 (2009-05-22)
Etretinate combined with low-dose prednisolone for an aged patient with adult T-cell leukaemia/lymphoma.
K Senba-Nakata et al.
Clinical and experimental dermatology, 35(4), e153-e154 (2009-11-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service