Skip to Content
Merck
All Photos(1)

Key Documents

Z777039

SRT® SEC-100 (5 µm) HPLC Columns

L × I.D. 30 cm × 7.8 mm, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

SRT® SEC-100 Gel Filtration Column, 5 μm, L × I.D. 30 cm × 7.8 mm, stainless steel

material

stainless steel

Agency

suitable for USP L59

product line

SRT®

mol wt

100-100,000

manufacturer/tradename

Sepax 215100-7830

packaging

1 ea of

parameter

20 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

5 μm

pore size

100 Å

pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Sepax SRT SEC columns feature a hydrophilic surface layer that effectively shields the highly porous, five micron silica particles from interacting with your sample. SRT columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.

SRT User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SRT is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service