Skip to Content
Merck
All Photos(1)

Key Documents

CRM48140

Supelco

EPA 501/601 Trihalomethanes Calibration Mix

certified reference material, TraceCERT®, 2000 μg/mL in methanol, ampule of 1 mL

Synonym(s):

THMs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 601,501.3
suitable for EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EPA 501/601 Trihalomethanes Calibration Mix is a multi-component solution which contains trihalomethanes EPA certified reference standards (CRMs) such as chloroform, bromodichloromethane, dibromochloromethane, and bromoform.
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

Application

EPA 501/601 Trihalomethanes Calibration Mix may be used as a CRM to determine the analytes such as chloroform, bromodichloromethane, dibromochloromethane, and bromoform in water samples by gas chromatography-mass spectrometry (GC-MS) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bromodichloromethane
    Bromoform
    Chloroform
    Dibromochloromethane

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Modeling the transport of neutral disinfection byproducts in forward osmosis: Roles of reverse salt flux
Xu J, et al.
Water Research, 185(1), 116255-116255 (2020)
Simultaneous Determination of Disinfection By-products in Water Samples from Advanced Membrane Treatments by Headspace Solid Phase Microextraction and Gas Chromatography-Mass Spectrometry
Martinez C, et al.
Analytical and Bioanalytical Chemistry Research, 1(1), 38-49 (2014)
Impact of chitosan and polyacrylamide on formation of carbonaceous and nitrogenous disinfection by-products
Li Z, et al.
Chemosphere, 178, 26-33 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service