Skip to Content
Merck
All Photos(1)

Key Documents

821455

TSKgel® ODS-100V (5 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

Product Name

TSKgel® ODS-100V HPLC Column, phase C18 (octadecyl), L × I.D. 15 cm × 4.6 mm, 5 μm particle size

material

stainless steel hardware

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh ODS-100V, 21455

extent of labeling

15% Carbon, endcapped with bifunctional dialkylsilane

parameter

0-100% organic solvent
1.0 mL/min standard flow rate
10-50 °C temperature
180 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

450 m2/g

matrix

silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® ODS-100V and TSKgel ODS-100Z columns incorporate the best-in-class surface properties to limit secondary interactions of basic, acidic and chelating compounds. Take advantage of the benefits of high column efficiency and symmetrical peak shapes to reduce your analysis and method development time! The ultra high purity Type B base silica contains negligible amounts of metal ion impurities. Combined with monomeric bonding chemistry, this silica makes the best general purpose Reversed Phase columns suitable for the most demanding separations in quality control as well as in research and development. TSKgel ODS-100V columns have a lower carbon content than ODS-100Z columns, making the bonded phase surface 100% water wettable and the preferred choice to retain polar metabolites.

Application

TSKgel® ODS-100V HPLC Column may be used for the separation of:
  • Docetaxel in human plasma during its determination by high performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) combined with electrospray ionization (ESI) and multiple reaction monitoring (MRM) detection.
  • Constituents of Chinese medicine Niu Huang Jie Du Pill during their analysis by HPLC combined with diode array detection (DAD).
  • Organic and inorganic selenium in diet supplements during the determination using HPLC-ESI-MS/MS with MRM detection.
  • Estrogenic compounds 8-prenylnaringenin, zearalenone, α- and β-zearalenol in beer during their identification using LC-ESI-MS with single ion monitoring (SIM) detection.
  • Lincomycin, spectinomycin and its impurities in pharmaceutical products during their analysis by HPLC combined with charged aerosol detection (CAD) as well as HPLC coupled to time-of flight mass spectrometer (TOFMS) with ESI.

Physical form

Shipped in acetonitrile.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous quantitation and identification of organic and inorganic selenium in diet supplements by liquid chromatography with tandem mass spectrometry.
Zembrzuska J, et al.
Food Chemistry, 142(3), 178-187 (2014)
A rapid and sensitive LC/ESI-MS/MS method for quantitative analysis of docetaxel in human plasma and its application to a pharmacokinetic study.
Yamaguchi H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 893, 157-161 (2012)
Qualitative and quantitative analysis of traditional Chinese medicine Niu Huang Jie Du Pill using ultra performance liquid chromatography coupled with tunable UV detector and rapid resolution liquid chromatography coupled with time-of-flight tandem mass spectrometry
Liang X, et al.
Journal of Pharmaceutical and Biomedical Analysis, 51(3), 565-571 (2010)
Development of high performance liquid chromatography methods with charged aerosol detection for the determination of lincomycin, spectinomycin and its impurities in pharmaceutical products
Stypulkowska K, et al.
Journal of Pharmaceutical and Biomedical Analysis, 112(1), 8-14 (2015)
Direct determination of the estrogenic compounds 8-prenylnaringenin, zearalenone, ?- and ?-zearalenol in beer by liquid chromatography-mass spectrometry.
Maragou NC, et al.
Journal of Chromatography A, 1202(1), 47-57 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service