Skip to Content
Merck
All Photos(1)

Key Documents

581401-U

Supelco

Ascentis® C8 (3 µm) HPLC Columns

L × I.D. 10 cm × 2.1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Ascentis® C8 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm

material

stainless steel column

Agency

suitable for USP L7

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

extent of labeling

15% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

10 cm × 2.1 mm

surface area

450 m2/g

surface coverage

4.0 μmol/m2

impurities

<5 ppm metals

matrix

fully porous particle
silica gel high purity, spherical

matrix active group

C8 (octyl) phase

particle size

3 μm

pore size

100 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The Ascentis family of columns is the fourth generation of HPLC column technology from Supelco scientists. Ascentis columns are bonded on high purity, 100 Angstrom silica including 3, 5, and 10 micron particle size. Columns are designed for small molecule applications and are scalable from micro columns (1.0 mm I.D.) to preparative dimensions (50 mm I.D.). The family includes C18, C8, Phenyl, Si and embedded polar group phase, RP-Amide.

The Ascentis C8 is a highly reproducible column with excellent selectivity towards polar compounds. The phase excells in highly aqueous mobile phases as well as high organic mobile phases.

Features and Benefits

  • Superior retention for hydrophobic molecules
  • Excellent peak shape
  • Low bleed LC-MS separations

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahmoud Hasan et al.
Journal of pharmaceutical and biomedical analysis, 100, 341-347 (2014-09-10)
Clodronate belongs to the class of bisphosphonates which are used for the treatment of bone disorders. Due to its high polarity it has a low and highly variable oral bioavailability which results in low plasma concentrations and requires sensitive bioanalytical
Ebenezer B Asafu-Adjaye et al.
Journal of pharmaceutical and biomedical analysis, 43(5), 1854-1859 (2007-02-16)
Gastrointestinal stability of venlafaxine was evaluated in vitro in simulated gastric (SGF) and intestinal (SIF) fluids using a stability indicating HPLC method. The method was validated using a 5 microm Ascentis C18 column (150 mm x 4.6 mm) and mobile
Luana Magalhães et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1008, 98-107 (2015-12-08)
Nucleoside triphosphate diphosphohydrolase (NTPDase) is an enzyme belonging to the apyrase family that participates in the hydrolysis of the nucleosides di- and triphosphate to the corresponding nucleoside monophosphate. This enzyme underlies the virulence of parasites such as Leishmania. Recently, an
Sreenivasa Rao Chitturi et al.
Journal of pharmaceutical and biomedical analysis, 55(1), 31-47 (2011-02-15)
This paper proposes a simple and selective RP-HPLC method for the determination of process impurities and degradation products (degradants) of atazanavir sulfate (ATV) drug substance. Chromatographic separation was achieved on Ascentis(®) Express C8, (150mm×4.6mm, 2.7μm) column thermostated at 30°C under
Ugandar Reddy Inugala et al.
Journal of chromatographic science, 51(5), 453-459 (2012-10-13)
This paper describes the development of a rapid, novel, stability-indicating gradient reversed-phase high-performance liquid chromatographic method and associated system suitability parameters for the analysis of naproxcinod in the presence of its related substances and degradents using a quality-by-design approach. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service