Documents
57171
Supelco
ENVI-18 DSK SPE Disk
diam. 47 mm, pk of 24
About This Item
UNSPSC Code:
41115712
Agency
suitable for EPA 1613
Quality Level
product line
ENVI
packaging
pk of 24
technique(s)
solid phase extraction (SPE): suitable
diam.
47 mm
application(s)
food and beverages
separation technique
reversed phase
General description
Retention Mechanism: Reversed-phase
Sample Matrix Compatibility: Aqueous solutions (drinking water)
  • The SPE membrane equivalents of ENVI-18 and ENVI-8 packed bed SPE sorbents
  • Porous glass fiber membranes embedded with C18 or C8 modified silica particles
  • Provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples
  • Typical applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides
Legal Information
ENVI is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)