Skip to Content
Merck
All Photos(1)

Key Documents

57018

Supelco

Supelclean LC-SCX SPE Tube

bed wt. 500 mg, volume 3 mL, pkg of 54 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712

material

polypropylene tube

product line

Supelclean

feature

endcapped

composition

bed wt., 500 mg

packaging

pkg of 54 ea

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

matrix active group

SCX phase
SCX

diameter

40-45 μm

pore size

60 Å

application(s)

food and beverages

separation technique

ion exchange
ion exchange

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Supelclean LC-SCX SPE Tube may be used for the solid-phase extraction of:
  • Cimaterol, salbutamol, terbutaline and ractopamine in feed samples prior to their determination by ultra-performance liquid chromatography-fluorescence detection (UPLC-FLD).
  • Adulterants in slimming functional foods prior to their quantification by gas chromatography-electron impact-tandem mass spectrometry (GC-EI-MS/MS).

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A GC-EI-MS-MS Method for Simultaneous Determination of Seven Adulterants in Slimming Functional Foods.
Li Y, et al.
Journal of Chromatographic Science, 50(10), 928-933 (2012)
Simultaneous determination of cimaterol, salbutamol, terbutaline and ractopamine in feed by SPE coupled to UPLC.
Qu CH, et al.
Chromatographia, 73(3-4), 243-249 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service