Skip to Content
Merck
All Photos(1)

Key Documents

569232-U

Supelco

Discovery® C18 (5 µm) HPLC Columns

L × I.D. 12.5 cm × 4.6 mm, HPLC Column

Synonym(s):

Discovery RP18 HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

12% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

12.5 cm × 4.6 mm

surface area

300 m2/g

surface coverage

3 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

180 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Use Discovery® C18 for any method that specifies a C18. The exceptional peak shape, reproducibility, and stability make it the column of choice for all C18 methods from demanding to routine.

Application


  • Rat Pharmacokinetics and In Vitro Metabolite Identification of KM-819, a Parkinson′s Disease Candidate, Using LC-MS/MS and LC-HRMS.: This study underscores the utility of theDiscovery® C18 HPLC Column in pharmacokinetic profiling and metabolite identification of a novel therapeutic agent, demonstrating its critical role in the development of Parkinson′s disease treatments (Choi et al., 2024).

  • Determination of 2,4-dimethylhydroxybenzene by chromatographic methods in forensic toxicological research of biological material.: Highlights the application of the Discovery® C18 HPLC Column in forensic toxicology, enhancing the detection and quantification of toxic substances within biological samples, vital for legal and medical investigations (Chernova et al., 2024).

  • High-throughput microfluidic chip with silica gel-C18 channels for cyclotide separation.: Illustrates the integration of the Discovery® C18 HPLC Column in a microfluidic setting for efficient and high-throughput separation of cyclotides, offering advancements in peptide and protein analysis (Ebrahimi et al., 2023).

  • Priestia megaterium Metabolism: Isolation, Identification of Naringenin Analogues and Genes Elevated Associated with Nanoparticle Intervention.: This research utilizes the Discovery® C18 HPLC Column for the advanced study of microbial metabolism under nanoparticle intervention, pushing the boundaries of biotechnological and pharmaceutical research (Al-Theyab et al., 2023).

  • A practical strategy enabling more reliable identification of ginsenosides from Panax quinquefolius flower by dimension-enhanced liquid chromatography/mass spectrometry and quantitative structure-retention relationship-based retention behavior prediction.: Demonstrates the versatility of the Discovery® C18 HPLC Column in complex botanical analyses, particularly in enhancing the reliability of ginsenoside identification, crucial for the development of herbal medicinal products (Sun et al., 2023).

Features and Benefits

  • Excellent reproducibility
  • Exceptional peak shape for basic and acidic analytes
  • Stable, low-bleed LC-MS separations
  • Separation of peptides and small proteins
  • Lower hydrophobicity than many comparable C18 columns, providing faster analysis

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Nageswara Rao et al.
Biomedical chromatography : BMC, 23(11), 1145-1150 (2009-05-16)
A highly sensitive and selective on-line two-dimensional reversed-phase liquid chromatography/electrospray ionization-tandem mass spectrometry (2D-LC-ESI/MS/MS) method was developed and validated to determine rifaximin in rat serum by direct injection. The 2D-LC-ESI/MS/MS system consisted of a restricted access media column for trapping
Yoshinori Masukawa et al.
Journal of lipid research, 49(7), 1466-1476 (2008-03-25)
Ceramides (CERs) in human stratum corneum (SC) play physicochemical roles in determining barrier and water-holding functions of the skin, and specific species might be closely related to the regulation of keratinization, together with other CER-related lipids. Structures of those diverse
Pei Yuan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(42), 11319-11325 (2009-09-18)
This study systematically investigates periodic mesoporous organosilicas (PMOs) with controlled helical and concentric circular (CC) pore architectures prepared through a basic-catalyzed sol-gel process by using an achiral cationic surfactant trimethyloctadecylammonium bromide (C(18)TAB) as a structure-directing agent, perfluorooctanoic acid (PFOA) as
Pilar Heredia-Middleton et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 149(4), 552-556 (2008-02-01)
Heat shock protein 70 (HSP70, 70 kDa) is the most commonly expressed protein in response to thermal stress. The extent of its expression is associated with differences in environmental temperatures. We investigated the heat shock response in red blood cells
Xiaofang Hou et al.
Journal of separation science, 36(4), 706-712 (2013-02-13)
Most of the anti-breast cancer drugs are often limited owing to drug resistance and serious adverse reactions. Therefore, development of more targeted and low toxic drugs from traditional Chinese medicines for breast cancer are needed. At the same time, establishment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service