Skip to Content
Merck
All Photos(1)

Key Documents

567576-U

Supelco

Discovery® HS F5 (5 µm) HPLC Columns

L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge, pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® HS F5 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

material

stainless steel column

Agency

suitable for USP L43

description

Supelguard Cartridge

product line

Discovery®

feature

endcapped

packaging

pkg of 2 ea

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 4 mm

surface area

300 m2/g

matrix

fully porous particle

matrix active group

PFP (pentafluorophenyl) phase

particle size

5 μm

pore size

120 Å

operating pH

2-8

application(s)

food and beverages

compatibility

use to protect Discovery HS F5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Packaging

For 4.0mm I.D. and 4.6mm I.D. analytical columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umberto M Battisti et al.
Journal of chromatography. A, 1443, 152-161 (2016-03-30)
A "heart-cut" two-dimensional achiral-chiral liquid chromatography triple-quadrupole mass spectrometry method (LC-LC-MS/MS) was developed and coupled to in vivo cerebral microdialysis to evaluate the brain response to the chiral compound (±)-7-chloro-5-(3-furanyl)-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-1,1-dioxide ((±)-1), a potent positive allosteric modulator (PAM) of AMPA receptor.
Jennie Wang et al.
Journal of pharmaceutical and biomedical analysis, 124, 267-273 (2016-03-13)
An HPLC method for the assay of a DNA topoisomerase inhibitor, LMP776 (NSC 725776), has been developed and validated. The stress testing of LMP776 was carried out in accordance with International Conference on Harmonization (ICH) guidelines Q1A (R2) under acidic
Lucie Havlíková et al.
Talanta, 76(3), 597-601 (2008-07-01)
A pentafluorophenylpropyl (PFPP) stationary phase was for the first time tested for the simultaneous determination of triamcinolone acetonide, its degradation product triamcinolone and two preservatives, methylparaben, and propylparaben. A new simple isocratic reversed phase HPLC method with UV detection, using
Jana Maláková et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 853(1-2), 265-274 (2007-04-10)
Galantamine, an alkaloid isolated from the bulbs and flowers of Caucasian snowdrop (Galanthus woronowii, Amaryllidaceae) and related species, is employed in human medicine for the treatment of various neuromuscular and neurodegenerative diseases. After the administration, the products of oxidative biotransformation
C Desmortreux et al.
Journal of chromatography. A, 1216(42), 7088-7095 (2009-09-15)
Separation of furocoumarins has become of a great interest for cosmetic industry and human health, since the recent directive of the European Union. Furocoumarins are a class of compounds presenting varied substituents linked mainly in two positions to an identical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service