Skip to Content
Merck
All Photos(1)

Key Documents

55442-U

Supelco

Supel QuE Verde QuEChERS tube

Tube, centrifuge tube volume 15 mL, pack of 50 ea

Synonym(s):

Supel QuE, QuEChERS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

product name

Supel QuE Verde, Tube, centrifuge tube volume 15 mL, pack of 50 ea

material

polypropylene tube

Quality Level

product line

Supel

composition

magnesium sulfate, 1200 mg
Supelclean ENVI-Carb Y, 80 mg
Supelclean PSA, 400 mg
Z-Sep+, 480 mg

packaging

pack of 50 ea

technique(s)

QuEChERS: suitable

centrifuge tube volume

15 mL

matrix active group

PSA phase
carbon phase
zirconia-based phase

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.

Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.

Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.

Suitability

Designed to provide high recovery all pesticides, including problematic planar pesticides, in samples containing high levels of chlorophyll (e.g. cannabis, spinach, lettuce, oregano, etc.). Ideal for cleanup of extract after initial food extraction and phase partitioning.

Legal Information

AOAC is a registered trademark of AOAC International
ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
Supel is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service