55408-U
Supel™ QuE QuEChERS tube
Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Synonym(s):
QuEChERS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115712
NACRES:
NB.21
Recommended Products
Product Name
Supel™ QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
material
polypropylene tube
Quality Level
product line
Supel™
composition
Z-Sep+, 75 mg
packaging
pack of 100 ea
technique(s)
QuEChERS: suitable
centrifuge tube volume
2 mL
matrix active group
zirconia-based phase
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
General description
Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.
Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.
Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.
Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.
Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.
Other Notes
Supel™ QuE Z-Sep products enhance sample cleanup for complex matrices by effectively removing more fat and color from sample extracts than traditional phases for QuEChERS methods. By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS and GC-MS methods. This proprietary technology can replace C18 and PSA in your current methods without additional method development.
Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.
Z-Sep/C18 is recommended for cleanup of samples containing less than 15% fat.
Z-Sep is recommended for cleanup of samples prior to analysis of hydrophobic analytes.
Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.
Z-Sep/C18 is recommended for cleanup of samples containing less than 15% fat.
Z-Sep is recommended for cleanup of samples prior to analysis of hydrophobic analytes.
Legal Information
Certain uses of this product may be covered by the claims of one or more pending patent applications. For licensing inquiries please contact [email protected]
AOAC is a registered trademark of AOAC International
Supel is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
An overview of analytical methods and occurrence of benzotriazoles, benzothiazoles and benzenesulfonamides in the environment
Herrero, P., et al.
TrAC, Trends in Analytical Chemistry, 62, 46-55 (2014)
Anna Laura Capriotti et al.
Journal of chromatography. A, 1343, 69-78 (2014-04-22)
A sensitive, simple and rapid method for the simultaneous determination of 19 mycotoxins in biscuits (a dry matrix containing cereals and egg) has been developed using high performance liquid chromatography coupled to tandem mass spectrometry with electrospray source working in
Determination of toxic substances, pesticides and mycotoxins, in ginkgo biloba nutraceutical products by liquid chromatography Orbitrap-mass spectrometry
Martinez-Dominguez, G., et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 118, 124-130 (2015)
Ana Lozano et al.
Talanta, 118, 68-83 (2013-11-28)
Two sorbents containing ZrO₂ (Z-Sep and Z-Sep+) were tested as a d-SPE clean-up in combination with the QuEChERS and ethyl acetate multiresidue method in the pesticide residues extraction in avocado. All extracts were analysed using gas chromatography coupled with a
Jonatan V Dias et al.
Journal of chromatography. A, 1462, 8-18 (2016-08-11)
The goal of this work was to evaluate the efficiency of several sorbents on removal fats from edible oils (olive, soya and sunflower) during the clean-up step for posterior determination of 165 pesticides by UHPLC-QqQ-MS/MS system. The extraction procedure employed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service