47266
Sugar Alcohol Kit
(individually packaged in quantities indicated), analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
analytical standard
CofA
current certificate can be downloaded
packaging
pkg of 1 kit
concentration
(individually packaged in quantities indicated)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
storage temp.
-10 to -25°C
General description
Sugar Alcohol Kit contains individually packaged standards in the quantities listed. The sugar alcohols are prepared, tested, and individually packaged using rigorous manufacturing procedures.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Kit Components Only
Product No.
Description
- D-(+)Arabitol 500 mg
- Dulcitol (Galactitol) 500 mg
- iso-Erythritol 500 mg
- Glycerol 500 mg
- Maltitol 500 mg
- D-Mannitol 500 mg
- Ribitol (Adonitol) 500 mg
- D-Sorbitol 500 mg
- Xylitol 500 mg
See All (9)
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service