Skip to Content
Merck
All Photos(1)

Key Documents

44807

Supelco

Aroclor 1248 solution

certified reference material, 1000 μg/mL in isooctane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8082

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Clc1cc(cc(c1)c2cc(cc(c2)Cl)Cl)Cl

InChI

1S/C12H6Cl4/c13-9-1-7(2-10(14)5-9)8-3-11(15)6-12(16)4-8/h1-6H

InChI key

UTMWFJSRHLYRPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jongseol Kim et al.
Journal of microbiology and biotechnology, 18(10), 1701-1708 (2008-10-29)
Previous investigations showed that three classes of haloaromatic compounds (HACs; chlorobenzoates, chlorophenols, and chlorobenzenes) enhanced the reductive dechlorination of Aroclor 1248, judging from the overall extent of reduction in Cl atoms on the biphenyl. In the present study, we further
Mercedes Fernández Santiago et al.
Toxicology letters, 163(2), 91-100 (2005-11-03)
We have demonstrated previously [Pérez-Reyes, P.L., Sánchez-Alonso, J.A., López-Aparicio, P., Recio, M.N., Pérez-Albarsanz, M.A., 2001. Different molecular capacity in the induction of apoptosis by polychlorinated biphenyl congeners in rat renal tubular cell cultures. Biosci. Rep. 6, 765-778] that the polychlorinated
Jennifer E Low et al.
Environmental pollution (Barking, Essex : 1987), 159(3), 769-775 (2010-12-21)
Two cultivation techniques (i-pruning and ii-nodal adventitious root encouragement) were investigated for their ability to increase PCB phytoextraction by Cucurbita pepo ssp pepo cv. Howden (pumpkin) plants in situ at a contaminated industrial site in Ontario (Aroclor 1248, mean soil
Andres Martinez et al.
Environment international, 36(8), 849-854 (2009-03-10)
We report the results of the first intensive survey of polychlorinated biphenyls (PCBs) in the surficial sediment of the Indiana Harbor and Ship Canal (IHSC) in East Chicago, Indiana, a part of the Calumet River tributary of Lake Michigan that
Alisa Bronshtein et al.
Journal of agricultural and food chemistry, 60(17), 4235-4242 (2012-03-17)
Development of a multianalyte enzyme-linked immunosorbent assay (ELISA) for detection of permethrin and aroclors 1248 or 1254 and implementation of the assay for analysis of soil/sediment samples are described. The feasibility of using the multianalyte ELISA to monitor aroclors 1254

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service