Skip to Content
Merck
All Photos(1)

Key Documents

442811

Supelco

trans-Nonachlor

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

ClC21C3C(C(C2(Cl)Cl)(C(=C1Cl)Cl)Cl)C(C(C3Cl)Cl)Cl

InChI

1S/C10H5Cl9/c11-3-1-2(4(12)5(3)13)9(17)7(15)6(14)8(1,16)10(9,18)19/h1-5H

InChI key

OCHOKXCPKDPNQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohui Xu et al.
Environmental health perspectives, 118(1), 60-66 (2010-01-09)
Organochlorine (OC) pesticides are a group of environmental endocrine disruptors that may be associated with an increased risk for hormone-related cancers including cancers of the breast and prostate. However, epidemiologic evidence is limited and inconsistent. We used 1999-2004 National Health
Genevieve Bondy et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 42(6), 1015-1027 (2004-04-28)
The chlordane constituent trans-nonachlor and its metabolite oxychlordane are among the most persistent chlordane-related contaminants and are found in tissues and milk from humans ingesting diets high in Arctic marine mammal fat. Although chlordane is no longer registered in North
J S Lim et al.
International journal of obesity (2005), 35(5), 744-747 (2010-09-08)
There is emerging evidence that persistent organic pollutants (POPs) can increase the risk of various chronic diseases. As POPs mainly bioaccumulate in adipose tissue, weight change can affect serum concentrations of POPs. However, there are few population-based studies on effects
Mary S Wolff et al.
Environmental research, 97(2), 170-177 (2004-11-10)
Organochlorine compounds (OCs) have been found widely in human tissues. However, levels have been rapidly declining since their virtual ban in the 1970s. We measured 1,1'-dichloro-2,2'-bis(4-chlorophenyl)ethylene (DDE), polychlorinated biphenyls (PCBs), and trans-nonachlor (TN) in 194 pregnant women in New York
V M Chia et al.
International journal of andrology, 33(4), 588-596 (2009-07-25)
It has been hypothesized that the increased prevalence of testicular germ cell tumours (TGCT) may be attributable to endocrine disrupting chemicals, such as persistent organic pollutants (POPs); these may be modulated by hormone-metabolizing enzymes. Using data from 568 cases and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service