Skip to Content
Merck
All Photos(1)

Key Documents

30176

Supelco

TraceCERT®Multiresidue Pesticide Standard Mix #3

in acetonitrile, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

in acetonitrile
100 μg/mL each component in acetonitrile

application(s)

cannabis testing
food and beverages
(pesticide residue testing)

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Other Notes

21 components. When combined with other mixes, suitable for use in meeting California state requirements for pesticide testing of cannabis products
Check out our entire range of pesticide mixes for Cannabis testing

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Azoxystrobin
    Tebuconazole
    Myclobutanil
    Pyridaben
    Hexythiazox
    Spinosad
    N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine
    Fenpyroximate
    Trifloxystrobin
    Acequinocyl
    Bifenazate
    3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine
    Etoxazole
    Fenhexamid
    Flonicamid
    Propiconazole
    Abamectin
    Fludioxonil
    Boscalid
    Methyl (E)-α-(methoxyimino)-2-(2-methylphenoxymethyl)phenylacetate
    Dimethoate
    See All (21)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup - No data available

Flash Point(C)

2.0 °C - closed cup - No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct extraction with acetonitrile of hemp seed oil for the analysis of pesticides by using comprehensive two-dimensional gas chromatography-triple quadrupole mass spectrometry
Arena A, et al.
Journal of Chromatography A, 464432-464432 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service