29557-U
CGA-580 Gas Cylinder Pressure Regulator
2-stage high purity, parameter 0-100 psi
Synonym(s):
Pressure Regulator (He, Ar, N2)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41112500
NACRES:
SB.54
Recommended Products
parameter
0-100 psi
Looking for similar products? Visit Product Comparison Guide
General description
For use with helium, nitrogen, or argon gas cylinders.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service