24447
SGE Needles for RN syringes
volume 25-500 μL, needle size 25 ga, needle L × O.D. 50 mm × 0.50 mm
Synonym(s):
REPL.NEEDLE N25/500-5, PK5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
material
bevel tip stainless steel needle (point style 2)
needle size
25 ga
description
N25/500-5
replacement needles
packaging
pkg of 5 ea
manufacturer/tradename
SGE 038110
needle L × O.D.
50 mm × 0.50 mm
volume
25-500 μL
compatibility
for use with SGE 25R
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
RN syringes allow easy replacement of needles that get damaged in the field. The needle is seated at zero graduation mark.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service