23201
Space Saver storage racks
for use with 21-23 mm diameter vial (36 vials per rack), well opening 23.1 mm, pkg of 5 ea
Synonym(s):
VIAL RACK,21MM-23MM VIALS 5/CS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
red polypropylene rack
description
vial size: 15-22 mL
feature
positions 36 vials per rack
packaging
pkg of 5 ea
rack L × W × H
320 mm × 90 mm × 30 mm
well opening
23.1 mm
compatibility
for use with 21-23 mm diameter vial (36 vials per rack)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Economical and convenient storage for samples in your screw cap and crimp seal vials. Can be stacked when empty or full. Autoclavable, 120°C. Each case contains 5 racks.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service