20281
Florisil®
100-200 mesh, fine powder
Synonym(s):
Activated magnesium silicate, Magnesium silicate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
Recommended Products
grade
standard
form
fine powder
packaging
jar of 454 g
technique(s)
LPLC: suitable
surface area
289 m2/g
matrix
Magnesium silicate hydrate
matrix active group
magnesium silicate hydrate
particle size
100-200 mesh
74-149 μm
pore size
60 Å mean pore size
separation technique
hydrophilic interaction (HILIC)
SMILES string
[Mg++].[O-][Si]([O-])=O
InChI
1S/Mg.O3Si/c;1-4(2)3/q+2;-2
InChI key
FKHIFSZMMVMEQY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Florisil, adsorbent for the separation of lipids is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.
Application
Florisil was used:
- as column packing material in column chromatography
- as an adsorbent in gas chromatography with electron capture detector (GC/ECD), capillary column chromatography, adsorption column chromatography
- in clean-up procedures in gas chromatography-tandem mass spectrometry and GC-FID
Legal Information
Florisil is a registered trademark of U.S. Silica Company
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
An improved gas chromatographic method for the determination of perfluorocarbon tracers in the atmosphere
Lagomarsino, Raymond J.
Journal of Chromatographic Science, 34, 405-412 (1996)
P A de Paula Pereira et al.
Talanta, 49(2), 245-252 (2008-10-31)
A new sampling protocol was developed to determine methanol and ethanol in the gas phase, at low concentration levels, in urban atmospheres. The procedure involves collection of air samples (20.0-30.0 l) with three florisil cartridges connected in series, at a
The covalently bound flavin of Chromatium cytochrome c552. 1. Evidence for cysteine thiohemiacetal at the 8 alpha position.
W H Walker et al.
European journal of biochemistry, 48(2), 439-448 (1974-10-02)
Andrew C Singer et al.
Environmental toxicology and chemistry, 22(9), 1998-2004 (2003-09-10)
This study investigated the interactive effects of bioaugmentation, biostimulation, and the rhizosphere during remediation of Aroclor 1242-contaminated soil. Treatments were repeatedly augmented with polychlorinated bipheny (PCB)-degrading bacteria, inducers (carvone and salicylic acid), surfactant (sorbitan trioleate), minimal salts medium in a
R C Prados-Rosales et al.
Journal of chromatography. A, 993(1-2), 121-129 (2003-05-09)
A rapid analytical method for determination of organochlorine pesticide residues in sunflower seeds based on focused microwave-assisted Soxhlet extraction has been developed. The main factors affecting the extraction efficiency--namely microwave power, irradiation time, volume of extractant and number of cycles--were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service