Skip to Content
Merck
All Photos(1)

Key Documents

20051

Supelco

ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg

W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea

Synonym(s):

ORBO Amberlite XAD-2 Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W separators

Quality Level

Agency

NIOSH 2534,2537

product line

Amberlite
ORBO

composition

Bed A, 400 mg
Bed B, 200 mg

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 609

technique(s)

active air sampling: suitable

O.D. × L

8 mm × 110 mm

matrix

XAD-2

particle size

20-50 mesh

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

ORBO 609 Amberlite XAD®-2 (20/50), 400/200 mg sorbent tubes (W,W,W separators, O.D. × L 8 mm × 110 mm) contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Ørbo et al.
The Journal of steroid biochemistry and molecular biology, 113(1-2), 139-149 (2009-01-14)
Patients with endometrial hyperplasia representing preliminary stages of endometrial cancer have shown to respond to therapy in 100% of the cases when treated with levonorgestrel-impregnated intrauterine device. Anti-proliferative effect has also been reported after application of an anti-progestin impregnated intrauterine
Anne Beate Vereide et al.
Gynecologic oncology, 97(3), 740-750 (2005-05-12)
The aim of the study was to investigate apoptosis as a growth regulatory mechanism of gestagen in endometrial precancers and to compare differences in the apoptotic cascade after high and low dose gestagen regimens. Pre- and post-treatment paraffin-embedded endometrial hyperplasia
A Orbo et al.
International journal of oncology, 5(3), 619-625 (1994-09-01)
About 50% of the cases with invasive carcinoma of the uterine cervix occurs before menopause. Cyclic changes of sex steroids may therefore interfere with the ratio between extracellular levels of cGMP and cAMP (cGMP(ex)/cAMP(ex)). This ratio has been proposed as
H Stalsberg et al.
Virchows Archiv. A, Pathological anatomy and histopathology, 414(5), 415-422 (1989-01-01)
The autopsies of 12 victims from two snow avalanches in North-Norway are reported. Supportive evidence from non-autopsied and surviving victims is included. Consistent autopsy findings were prominent lung oedema, moderate cerebral oedema, extreme contraction of the left ventricle, petechiae in
Anne Ørbo et al.
Anticancer research, 35(12), 6401-6409 (2015-12-08)
To investigate if a levonorgestrel-impregnated intrauterine system (LNG-IUS) was more efficient compared to oral progestin in the clearance of the paired box 2 gene (PAX2) - and phosphatase and tensin homolog (PTEN)-null endometrial glands and assess the significance of PAX2-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service