13801
Metal Packed GC Column (General Configuration)
phase none, matrix 80/100 HayeSep Q support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
About This Item
Recommended Products
material
stainless steel column
parameter
0-275 °C temperature (isothermal or programmed)
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
matrix
80/100 HayeSep Q support
matrix active group
none phase
column type
packed GC
compatibility
for use with General Configuration
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service