Skip to Content
Merck
All Photos(1)

Key Documents

13028-U

Supelco

Carbon Adsorbent Sampler Kit

matrix CMS/SGPC Kit I

Synonym(s):

weak anion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
SB.54

product line

Carbosieve
Carboxen®
Graphsphere

form

solid

packaging

kit of 8 bottles

technique(s)

LPLC: suitable

matrix

CMS/SGPC Kit I
Carbon Molecular Sieve (CMS) and Spherical Graphitized Polymer Carbon (SGPC) adsorbents

matrix active group

carbon

particle size

(various mesh)

application(s)

sample preparation

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Choosing the right adsorbent or combination of adsorbents can be difficult:
  • Must choose one or more to retain specific analyte(s)
  • May be equally important to release analyte(s) during desorption

This convenient kit allows a cost-effective way to evaluate several of our specialty carbon adsorbents. Once you find an adsorbent or adsorbents that serve your purpose, we will work with you to either make your device for you, or to supply you bulk adsorbent (in a bottle, jar, or ampule; not packed inside a column or device).

For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon

Other Notes

Purchase of this kit is limited to two per customer per calendar year.

Footnote

Each bottle contains 5 g of a different material: GRAPHSPHERE 2016, 60/80 MESH 5G; CARBOXEN 569, 20/45 MESH 5G; CARBOXEN 1000, 60/80 MESH 5G;CARBOXEN 1012, 80/120 MESH 5G; CARBOXEN 1018, 60/80 MESH 5G;CARBOXEN 1021, 60/80 MESH 5G;CARBOSIEVE S-III, 60/80 MESH 5G; CARBOSIEVE G, 60/80 MESH 5G

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
Graphsphere is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • 20/45 Carboxen® 569 5 g

  • 60/80 Carboxen 1000 5 g

  • 80/120 Carboxen 1012 5 g

  • 60/80 Carboxen 1018 5 g

  • 60/80 Carboxen 1021 5 g

  • 60/80 Carbosieve G 5 g

  • 60/80 Carbosieve S-III 5 g

  • 60/80 Graphsphere 2016 5 g

See All (8)

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon molecular sieve gas separation membranes-I. Preparation and characterization based on polyimide precursors
Jones, Cheryl W., and William J. Koros
Carbon, 32, 1419-1425 (1994)
Carbon molecular sieve
Kaiser, R
Chromatographia, 3, 38-40 (1970)
Yukio Kemmochi et al.
Journal of chromatography. A, 977(2), 155-161 (2002-11-29)
2,3,7,8-Substituted polychlorinated dibenzo-p-dioxins/polychlorinated dibenzofurans (PCDD/Fs) and non-ortho-substituted polychlorinated biphenyls (PCBs) account for almost all of the total toxic equivalents (TEQ) in environmental samples. Activated carbon columns are used to fractionate the samples for GC-MS analysis or bioassay. Micropore-free surface-activated carbon
M Concejero et al.
Journal of chromatography. A, 917(1-2), 227-237 (2001-06-14)
Feasibility of several sorbents, Amoco PX-21, Carbosphere, Carbopack B and C and 2-(1-pyrenyl) ethyldimethylsilylated silica gel (PYE), for the fractionation of polychlorinated biphenyls, polychlorinated dibenzo-p-dioxins and furans was investigated. Selection was based on their suitability for quantitative isolation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service