Skip to Content
Merck
All Photos(1)

Key Documents

11982

Supelco

Tenax® Porous Polymer Adsorbent

matrix Tenax TA, 60-80 mesh, bottle of 10g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

Product Name

Tenax® Porous Polymer Adsorbent, matrix Tenax TA, 60-80 mesh, bottle of 10 g

Agency

suitable for EPA 601

Quality Level

form

solid

packaging

bottle of 10 g

parameter

350 °C temp. limit

surface area

~35 m2/g

matrix

Tenax TA

particle size

60-80 mesh

density

~0.25 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Tenax porous polymers, based on 2,6-diphenyl-p-phenylene oxide, is widely used as an adsorbent in both air collection and purge trap applications. Its unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Application

Tenax® Porous Polymer Adsorbent may be used for the adsorptive sampling of:
  • Volatile organic compounds (VOCs) in different microenvironments prior to their determination by short-path thermal desorption (SPTD) and gas chromatography-mass spectrometry (GC-MS).
  • C7-C26 hydrocarbons in ambient air samples and VOCs in workroom air samples prior to their analysis by TD followed by GC analysis.

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Water Analysis.
Nollet LM and De Gelder LSP.
Technology & Engineering, 625-626 (2007)
Adsorbent materials commonly used in air analysis for adsorptive enrichment and thermal desorption of volatile organic compounds.
Dettmer K and Engewald W
Analytical and Bioanalytical Chemistry, 373(6), 490-500 (2002)
Fire Investigation.
Daeid NN, et al.
Science, 142-143 (2004)
Stability of workroom air volatile organic compounds on solid adsorbents for thermal desorption gas chromatography.
Volden J, et al.
Analytica Chimica Acta, 530(2), 263-271 (2005)
Static Headspace-Gas Chromatography: Theory and Practice .
Kolb B and Ettre LS.
Science, 6-7 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service