Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1325

Sigma-Aldrich

Anti-β-Glucocerebrosidase Antibody, clone 2I15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2I15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 59.72 kDa
observed mol wt ~60 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey, porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2I15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects β-Glucocerebrosidase. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human β-Glucocerebrosidase.

Application

Quality Control Testing

Evaluated by Western Blotting in LNCap cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected β-Glucocerebrosidase in LNCap cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected β-Glucocerebrosidase in HeLa cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected β-Glucocerebrosidase in human liver and human pancreas tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected β-Glucocerebrosidase in LNCaP cells.

Affinity Binding Assay: A representative lot of this antibody bound β-Glucocerebrosidase peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-β-Glucocerebrosidase, clone 2I15 ZooMAb®, Cat. No. ZRB1325, is a recombinant Rabbit monoclonal antibody that detects β-Glucocerebrosidase and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry & Western Blotting.

Target description

Lysosomal acid glucosylceramidase (UniProt: P04062; also known as EC:3.2.1.45; Lysosomal acid GCase, Acid beta-glucosidase, Alglucerase, Beta-glucocerebrosidase, Beta-GC, Cholesterol glucosyltransferase, SGTase, Cholesteryl-beta-glucosidase, D-glucosyl-N-acylsphingosine glucohydrolase, Imiglucerase) is encoded by the GBA (also known as GC, GLUC) gene (Gene ID: 2629) in human. β-Glucocerebrosidase is a peripheral membrane glycoprotein that catalyzes the hydrolysis of glucosylceramide/GlcCer into free ceramide and glucose in the lysosomal compartment. It is shown to be maximally active at pH 5.5. Within the lysosome, it remains associated with the membrane, where it binds and degrades its substrate. Its interaction with saposin-C is reported to promote its membrane association. It plays a key role in the degradation of complex lipids and the turnover of cellular membranes. Through the production of ceramides, it participates in the PKC-activated salvage pathway of ceramide formation. It is also reported to plays a role in cholesterol metabolism and can catalyze the glucosylation of cholesterol, through a transglucosylation reaction that transfers glucose from glucosylceramide to cholesterol. Mutations in GBA gene are shown to cause Gaucher disease, a lysosomal storage disease characterized by accumulation of glucosylceramide in the reticulo-endothelial system. GBA mutations are also shown to be the strongest known risk factor for the development of Parkinson s disease. Five isoforms of β-Glucocerebrosidase have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service