Key Documents
Z743324
Breathe-EASY® petri dish strips, gas permeable polyurethane membrane
L × W 11.125 in. × 0.625 in.
Synonym(s):
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 100 ea
manufacturer/tradename
Diversified Biotech BEMP-1000
L × W
11.125 in. × 0.625 in.
General description
Breathe-Easy® breathable strips for sealing petri and cell culture dishes. Allows the exchange of oxygen and carbon dioxide for consistant cellular growth.
Legal Information
Breathe-Easy is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Questions
  1. Are these transparent? Or opaque/colored?
    1 answer
    1. These petri dish sealing strips are opaque. Please see the image below.
      Helpful?