Key Documents
Z694614
Nanofiber multiwell plate inserts, with aligned nanofibers
for 12 well plates
Synonym(s):
3D, 3D Cell Culture, 3d plates, tissue culture plates
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
Pricing and availability is not currently available.
material
transparent polycaprolactone (PCL)
transparent wells
sterility
sterile; electron beam irradiated
sterile
packaging
pack of 12 ea
manufacturer/tradename
Nanofiber Solutions 121202
size
12 wells
pore size
700 nm (Fiber spacing)
suitability
suitable for (Tissue Engineering, Cancer Research)
binding type
(plasma surface treated)
General description
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility.
The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining.
Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Which is the diameter of the nanofibers? How many nanofibers are in each slide?
    1 answer
    1. The diameter of the nanofibers is 700 nm. The exact number of nanofibers has not been documented. The thickness of the bottom of the plate is approximately 20 microns.
      Helpful?