Key Documents
Z331643
BRAND® Erlenmeyer flask, wide-neck, screw cap, PP
capacity 500 mL
Synonym(s):
conical flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
flat bottom
polypropylene
wide-neck
Joint
joint ST/NS45/40
joint neck GL52
sterility
non-sterile
feature
closure type Screw cap
with cap
packaging
case of 6
manufacturer/tradename
BRAND 93180
O.D. × H
108 mm × 180 mm
capacity
500 mL
General description
High clarity. With graduations. Threaded neck with standard ground socket.
Other Notes
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)