Skip to Content
Merck
All Photos(1)

Key Documents

Z328510

BRAND® Transferpettor caps

volume 100-200 μL

Synonym(s):

Pipettes, Piston-operated pipettes, Positive displacement principle, Repair set, Transferpettor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
100-200 μL
sterility:
non-sterile
feature:
color-coded blue/red
DE-M marking
packaging:
pack of 50 ea
manufacturer/tradename:
BRAND 701910
BRAND

material

glass

sterility

non-sterile

feature

color-coded blue/red
DE-M marking

packaging

pack of 50 ea

manufacturer/tradename

BRAND 701910
BRAND

maximum volume

200 μL

volume

100-200 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

BR701910 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service