Skip to Content
Merck
All Photos(2)

Key Documents

Y1626

Sigma-Aldrich

Select Yeast Extract, EZMix powder

for use in microbial growth medium

Synonym(s):

Select Yeast Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

grade

for molecular biology

Quality Level

sterility

non-sterile

form

powder

technique(s)

microbiological culture: suitable

application(s)

microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

SMILES string

OC(=O)CC1c2c3c(ccc2c4c1cccc4)cccc3

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Yeast extract is the water-soluble portion of autolyzed yeast, often used in culture media. Yeast extract contains a mixture of amino acids, peptides, water soluble vitamins and carbohydrates.

Application

For general bacteriological use with a variety of microorganisms.
SelectYeast Extract, EZMix powder has been used as a component of synthetic municipal wastewater fed to sequencing batch reactor (SBR) for biomass production. It has also been used as a supplement to super broth for the transformation of 5α-competent E. coli cells.
It is also suitable for use as nutritional source in microbial media.
Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Features and Benefits

Select Yeast Extract EZMix powder provides:
  • Granulated, dust-free format for safer handling and faster mixing
  • Easy scale-up using larger package sizes

Other Notes

Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Legal Information

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jayanta Haldar et al.
Nature protocols, 2(10), 2412-2417 (2007-10-20)
Protocols for the synthesis of the microbicidal polycation N,N-dodecyl,methyl-polyethylenimine and coating (painting) of glass slides with this polycation's butanol solution are described. Subsequently detailed are the procedures for validating that the resultant coated slides are essentially 100% lethal to the
F Morgan-Sagastume et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 69(1), 177-184 (2014-01-18)
Polyhydroxyalkanoates (PHAs) are biodegradable polyesters with comparable properties to some petroleum-based polyolefins. PHA production can be achieved in open, mixed microbial cultures and thereby coupled to wastewater and solid residual treatment. In this context, waste organic matter is utilised as
Artemis Kosta et al.
Methods in molecular biology (Clifton, N.J.), 346, 535-550 (2006-09-08)
In this chapter, we describe how to conveniently demonstrate, assess, and study cell death in Dictyostelium through simple cell culture, clonogenic tests, and photonic (with the help of staining techniques) and electronic microscopy. Cell death can be convniently generated using
Porter M Hall et al.
Nature structural & molecular biology, 29(12), 1217-1227 (2022-12-06)
CRISPR (clustered regularly interspaced short palindromic repeats) utility relies on a stable Cas effector complex binding to its target site. However, a Cas complex bound to DNA may be removed by motor proteins carrying out host processes and the mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service