Skip to Content
Merck
All Photos(1)

Documents

WS16

Sigma-Aldrich

Wright Stain, Modified

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
eCl@ss:
42039003
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.3%

pH

6.75-6.95 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Wright stain is a polychromatic dye, used for microscopic examination of blood to identify different cell types.

Application

Wright Stain, Modified has been used in:
  • Phagocytosis assay and puberty.
  • Estrous cycle examinations.
  • Staining, to differentiate neutrophils.
Popular hematology stain used for differentially staining the cellular elements of blood. For dip, rack, and batch staining techniques.

Other Notes

Wright stain, 0.3%, buffered at pH 6.8 in methanol. Contains stabilizers and surfactant.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Loss of magel2, a candidate gene for features of Prader-Willi syndrome, impairs reproductive function in mice
Mercer, Rebecca E and Wevrick, Rachel
PLoS ONE, 4(1), e4291-e4291 (2009)
Arginine and glutamine supplementation to culture media improves the performance of various channel catfish immune cells
Pohlenz C, et al.
Fish & Shellfish Immunology, 32(5) (2012)
Hematology: Clinical Principles and Applications (2007)
Fa-Zhao Wang et al.
Experimental and therapeutic medicine, 14(6), 6183-6192 (2017-12-19)
In the present study, the molecular mechanisms involved in the α-tomatine-induced apoptosis in human glioblastoma cell lines A172 and U-118 MG were investigated. Wright staining and ApopTag assays were conducted to confirm the apoptosis induced by α-tomatine treatment. Fura-2 assay
A potential mechanism for ADC-induced neutropenia: role of neutrophils in their own demise
Zhao H, et al.
Molecular Cancer Therapeutics, 4(1) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service