WHA10426892
Whatman® gel blotting paper, Grade GB003
GB003 blotting sheets, 58 x 60 cm, 50/pk
Synonym(s):
Western blotting paper, Z613886, blotting paper, protein blotting paper
About This Item
material
sheets
feature
Grade GB003
packaging
pack of 50
manufacturer/tradename
Whatman 10426892
Whatman Article No. 28415422 (US reference)
L × W
58 cm × 60 cm
thickness
0.8 mm
suitability
suitable for (lysis/denaturation of colony or plaque lifts and Western blots)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
GB003
A general purpose blotting paper (0.8 mm) made from pure raw materials with a high absorbency used as a membrane gel support. A thick paper recommended for the lysis/denaturation of colony or plaque lifts and Western blots.
Features and Benefits
- Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind. Ensures that no contamination will occur during the transfer steps.
- Manufactured and tested specifically for chromatographic and blotting techniques. This ensures the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting.
- Whatman 3MM Chr is considered the industry standard for blotting procedures.
- Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes. Allows "out-of-the-box" usage and eliminates sheet-to-sheet variations.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service