Skip to Content
Merck
All Photos(2)

Key Documents

WH0057526M4

Sigma-Aldrich

Monoclonal Anti-PCDH19 antibody produced in mouse

clone 2G10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp686P1843, Anti-KIAA1313, Anti-protocadherin 19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
2G10, monoclonal
species reactivity:
human
citations:
3
technique(s):
indirect ELISA: suitable
western blot: 1-5 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCDH19(57526)

Related Categories

General description

Protocadherin 19 (PCDH19) is part of the d-protocadherin superfamily. It is expressed in the brain. The gene encoding this calcium-dependent cell adhesion protein is localized on human chromosome Xq22.1.

Immunogen

PCDH19 (NP_065817, 241 a.a. ~ 340 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RLVPRDVEETDKMNVVSCSSLTSSLNYFDYHQQTLPLGCRRSESTFLNVENQNTRNTSANHIYHHSFNSQGPQQPDLIINGAPLPETENYSFDSNYVNSR

Biochem/physiol Actions

Protocadherin 19 (PCDH19) has been associated with female-specific epilepsy and an X-linked model of neurological disease. It has a role in neuronal organization and migration. PCDH19 is also involved in cell-cell and cell-matrix adhesion.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The clinical spectrum of female epilepsy patients with PCDH19 mutations in a Chinese population
A. Liu
Clinical Genetics, 91(1) (2017)
Male patients affected by mosaic PCDH19 mutations: five new cases
I.M. de Lange
Neurogenetics (2017)
Characterizing PCDH19 in human induced pluripotent stem cells (iPSCs) and iPSC-derived developing neurons: emerging role of a protein involved in controlling polarity during neurogenesis
Claudia Compagnucci
Oncotarget, 6(29), 26804-26813 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service