Skip to Content
Merck
All Photos(3)

Key Documents

WH0004288M1

Sigma-Aldrich

Monoclonal Anti-MKI67 antibody produced in mouse

clone 7B8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-KIA, Anti-Ki67

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7B8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKI67(4288)

General description

Ki-67 (antigen KI-67) has a gene size of 29,545 bp and it consists of 15 exons and 14 introns. It has a minus strand orientation. This gene is located on human chromosome 10q26.2.
This gene encodes a nuclear protein that is associated with and may be necessary for cellular proliferation. Alternatively spliced transcript variants have been described. A related pseudogene exists on chromosome X. (provided by RefSeq)

Immunogen

MKI67 (NP_002408, 3157 a.a. ~ 3256 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RSARQNESSQPKVAEESGGQKSAKVLMQNQKGKGEAGNSDSMCLRSRKTKSQPAASTLESKSVQRVTRSVKRCAENPKKAEDNVCVKKIRTRSHRDSEDI

Application

Monoclonal Anti-MKI67 antibody has been used in immunohistochemistry (IHC).

Biochem/physiol Actions

Ki-67 (antigen KI-67) protein acts as a cellular marker for proliferation. This protein plays a major role in the maintenance and regulation of the cell division cycle. Ki-67, a part of the mitotic chromosome periphery, function as a biological surfactant to maintain individual mitotic chromosomes dispersed in the cytoplasm after nuclear envelope disassembly.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service