Skip to Content
Merck
All Photos(1)

Key Documents

W1020

Sigma-Aldrich

W146 hydrate

≥98% (HPLC)

Synonym(s):

ML056, R-3-amino-4-(3-hexylphenylamino)-4-oxobutylphosphonic acid hydrate, [(3R)-3-amino-4-[(3-hexylphenyl)amino]-4-oxobutyl]-phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27N2O4P · xH2O
CAS Number:
Molecular Weight:
342.37 (anhydrous basis)
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

methanol: soluble (with 0.1% TFA)

originator

Novartis

storage temp.

2-8°C

SMILES string

O.CCCCCCc1cccc(NC(=O)[C@H](N)CCP(O)(O)=O)c1

InChI

1S/C16H27N2O4P.H2O/c1-2-3-4-5-7-13-8-6-9-14(12-13)18-16(19)15(17)10-11-23(20,21)22;/h6,8-9,12,15H,2-5,7,10-11,17H2,1H3,(H,18,19)(H2,20,21,22);1H2/t15-;/m1./s1

InChI key

DQKSZHVSROOTFY-XFULWGLBSA-N

Related Categories

General description

W146 hydrate functions as an orthosteric antagonist. It is associated with blood lymphopenia and lung edema in mice.

Application

W146 hydrate has been used to assess vascular permeability in mice.

Biochem/physiol Actions

Potent S1P(1) competitive antagonist; Ki = 10-77 nM.

Features and Benefits

This compound is featured on the Lysophospholipid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosine-1-Phosphate Signaling in Immunology and Infectious Diseases (2014)
The sphingosine-1-phosphate receptor-1 antagonist, W146, causes early and short-lasting peripheral blood lymphopenia in mice
Tarrason G, et al.
International Immunopharmacology, 11(11), 1773-1779 (2011)
Shahzad Nawaz Syed et al.
International journal of molecular sciences, 21(13) (2020-07-08)
Recent studies suggested an important contribution of sphingosine-1-phospate (S1P) signaling via its specific receptors (S1PRs) in the production of pro-inflammatory mediators such as Interleukin (IL)-1β in cancer and inflammation. In an inflammation-driven cancer setting, we previously reported that myeloid S1PR1
Impaired endothelial barrier function in apolipoprotein M-deficient mice is dependent on sphingosine-1-phosphate receptor 1
Christensen PM, et al.
Faseb Journal, 30(6), 2351-2359 (2016)
María V Simón et al.
Investigative ophthalmology & visual science, 56(10), 5808-5815 (2015-09-02)
Migration of Müller glial cells is enhanced in proliferative retinopathies, but the mechanisms involved are ill defined. Sphingosine-1-phosphate (S1P) is a bioactive sphingolipid synthesized by sphingosine kinase (SphK), which promotes proliferation, migration, and inflammation, acting as an intracellular mediator and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service