Skip to Content
Merck
All Photos(1)

Key Documents

V6258

Sigma-Aldrich

D-Val-Leu-Arg p-nitroanilide diacetate salt

≥95% (HPLC), suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38N8O5 · 2 C2H4O2
CAS Number:
Molecular Weight:
626.70
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

D-Val-Leu-Arg p-nitroanilide diacetate salt, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ~70%

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

CC(O)=O.CC(O)=O.N[C@H](C(C)C)C(N[C@@H](CC(C)C)C(N[C@@H](CCCNC(N)=N)C(NC1=CC=C([N+]([O-])=O)C=C1)=O)=O)=O

InChI

1S/C23H38N8O5.2C2H4O2/c1-13(2)12-18(30-22(34)19(24)14(3)4)21(33)29-17(6-5-11-27-23(25)26)20(32)28-15-7-9-16(10-8-15)31(35)36;2*1-2(3)4/h7-10,13-14,17-19H,5-6,11-12,24H2,1-4H3,(H,28,32)(H,29,33)(H,30,34)(H4,25,26,27);2*1H3,(H,3,4)/t17-,18-,19+;;/m0../s1

InChI key

WGVZWNUZAXIGKU-HYKWBEMSSA-N

Amino Acid Sequence

Val-Leu-Arg-pNA

Application

D-Val-Leu-Arg p-nitroanilide diacetate salt has been used as a substrate to determine the amidolytic activity of the fibrinolytic enzyme from Bacillus subtilis DC33. It has also been used as a substrate to determine the activity of fibrinolytic enzymes from the earthworm Eisenia fetida.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Quality

Salt content will vary.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Increase in ovarian kallikrein activity during ovulation in the gonadotrophin-primed immature rat.
Espey LL, Tanaka N, Winn V, Okamura H.
J. Reprod. Fert., 87, 503-508 (1989)
Purification and characterization of a fibrinolytic enzyme of Bacillus subtilis DC33, isolated from Chinese traditional Douchi
Cheng Tao Wang
Journal of Industrial Microbiology & Biotechnology, 33, 750-758 (2006)
I J Zeitlin et al.
The Journal of physiology, 543(Pt 1), 221-231 (2002-08-16)
It has been postulated that the mammary kinin system may play a role in modulating mammary blood flow. Until the present study, the local release of bradykinin (BK) or other kinin system constituents into the mammary vasculature had not been
József Dobó et al.
Journal of immunology (Baltimore, Md. : 1950), 183(2), 1207-1214 (2009-07-01)
Mannose-binding lectin (MBL)-associated serine protease (MASP)-1 is an abundant component of the lectin pathway of complement. The related enzyme, MASP-2 is capable of activating the complement cascade alone. Though the concentration of MASP-1 far exceeds that of MASP-2, only a
T L Miranda et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 28(5), 505-512 (1995-05-01)
Hydrolysis of D-valyl-L-leucyl-L-arginine p-nitroanilide (D-Val-Leu-Arg-Nan) at five different concentrations (10-20 microM) by human urinary kallikrein was studied in the absence and in the presence of increasing concentrations of basic pancreatic trypsin inhibitor (BPTI) (1.35-9.15 nM). The data indicate that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service