Skip to Content
Merck
All Photos(1)

Key Documents

V1014

Sigma-Aldrich

Anti-Vascular Endothelial Growth Factor Receptor 2 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Flk-1, Anti-VEGF R2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

flow cytometry: 3-10 μg/mL (106 cells)
immunohistochemistry: 5-15 μg/mL using cells and tissues
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Kdr(16542)

General description

Vascular endothelial growth factors receptors (VEGF Rs) are receptor tyrosine kinases that bind to VEGF that plays important role in embryonic vasculogenesis, angiogenesis and homeostasis. VEGF R2 has the ability to form heterodimers with VEGF R1 and VEGF R3. The binding of VEGF to VEGF R2 results in activation of tyrosine amino acids of the cytoplasmic tail of the receptor. This interaction results in the activation of MAPK, PI3K, PKC, FAK and Src kinase pathways. The biological roles of VEGF R2 include in epithelial cell growth, proliferation, survival, migration and vascular permeability and angiogenesis. Due to its role in promoting migration and angiogenesis, VEGF R2 has been implicated in breast and prostate cancer
Anti-Vascular Endothelial Growth Factor Receptor-2 recognizes mouse VEGF R2.

Specificity

The antibody has the ability to neutralize the biological activity of recombinant mouse VEGF R2. It demonstates approximately 40% cross-reactivity with recombinant human VEGF R2, 2% cross-reactivty with recombinant mouse VEGF R1, and no cross-reactivity with recombinant human VEGF R1 and VEGF R3.

Immunogen

purified recombinant mouse VEGF R2 extracellular domain expressed in NSO cells.

Application

Anti-Vascular Endothelial Growth Factor Receptor 2 antibody may be used in immunoblotting at a working concentration of 0.1-0.2 μg/ml. For flow cytometry the recommended concentration is 3-10 μg/ml. The immunohistochemistry of cells and tissues, a working concentration of 5-15 μg/ml may be used. For neutralization reactions, ND50 of 0.1-0.3 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanchun Guo et al.
Biochimica et biophysica acta, 1806(1), 108-121 (2010-05-14)
Investigations over the last decade have established the essential role of growth factors and their receptors during angiogenesis and carcinogenesis. The vascular endothelial growth factor receptor (VEGFR) family in mammals contains three members, VEGFR-1 (Flt-1), VEGFR-2 (KDR/Flk-1) and VEGFR-3 (Flt-4)
P Berger et al.
Swiss medical weekly, 141, w13318-w13318 (2011-12-20)
Vascular endothelial growth factors (VEGFs) regulate blood and lymph vessel formation through activation of the type V receptor tyrosine kinases VEGFR-1, -2 and -3. In addition, VEGFs interact with co-receptors such as neuropilins, integrins, semaphorins or heparansulfate glycosaminoglycans. Ligand binding
Rachel L Padget et al.
Mechanisms of development, 156, 8-19 (2019-02-24)
Blood vessel maturation, which is characterized by the investment of vascular smooth muscle cells (vSMCs) around developing blood vessels, begins when vessels remodel into a hierarchy of proximal arteries and proximal veins that branch into smaller distal capillaries. The ultimate
Nanbing Li-Villarreal et al.
Development (Cambridge, England), 149(7) (2022-03-18)
Establishing a functional circulatory system is required for post-implantation development during murine embryogenesis. Previous studies in loss-of-function mouse models showed that FOXO1, a Forkhead family transcription factor, is required for yolk sac (YS) vascular remodeling and survival beyond embryonic day
Hiroyuki Takahashi et al.
Clinical science (London, England : 1979), 109(3), 227-241 (2005-08-18)
The VEGF (vascular endothelial growth factor) family and its receptors are essential regulators of angiogenesis and vascular permeability. Currently, the VEGF family consists of VEGF-A, PlGF (placenta growth factor), VEGF-B, VEGF-C, VEGF-D, VEGF-E and snake venom VEGF. VEGF-A has at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service