Skip to Content
Merck
All Photos(1)

Key Documents

U5131

Sigma-Aldrich

Uroguanylin

≥90% (HPLC)

Synonym(s):

Asn-Asp-Asp-Cys-Glu-Leu-Cys-Val-Asn-Val-Ala-Cys-Thr-Gly-Cys-Leu [Disulfide bridges: 4–12, 7–15]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H102N18O26S4
CAS Number:
Molecular Weight:
1667.86
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Uroguanylin, ≥90% (HPLC)

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H]1CSSC[C@@H]2NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CSSC[C@H](NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC2=O)C(C)C)C(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N1)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](N)CC(N)=O)C(O)=O

InChI

1S/C64H102N18O26S4/c1-24(2)13-32-53(96)79-40-23-112-109-20-37(57(100)76-36(64(107)108)14-25(3)4)70-43(86)19-68-61(104)49(29(10)83)82-60(103)39(77-50(93)28(9)69-62(105)47(26(5)6)80-56(99)33(16-42(67)85)75-63(106)48(27(7)8)81-59(40)102)22-111-110-21-38(58(101)71-31(52(95)73-32)11-12-44(87)88)78-55(98)35(18-46(91)92)74-54(97)34(17-45(89)90)72-51(94)30(65)15-41(66)84/h24-40,47-49,83H,11-23,65H2,1-10H3,(H2,66,84)(H2,67,85)(H,68,104)(H,69,105)(H,70,86)(H,71,101)(H,72,94)(H,73,95)(H,74,97)(H,75,106)(H,76,100)(H,77,93)(H,78,98)(H,79,96)(H,80,99)(H,81,102)(H,82,103)(H,87,88)(H,89,90)(H,91,92)(H,107,108)/t28-,29+,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,47-,48-,49-/m0/s1

InChI key

VQZRYOAXGUCZMI-HWMZBCIHSA-N

Gene Information

human ... GUCA2B(2981)

Amino Acid Sequence

Asn-Asp-Asp-Cys-Glu-Leu-Cys-Val-Asn-Val-Ala-Cys-Thr-Gly-Cys-Leu [Disulfide bridges: 4–12, 7–15]

Biochem/physiol Actions

Stimulates intestinal guanylate cyclase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuleima Diaz et al.
Infection and immunity, 87(7) (2019-05-08)
Infection with enterotoxigenic Escherichia coli (ETEC) is a common cause of childhood diarrhea in low- and middle-income countries, as well as of diarrhea among travelers to these countries. In children, ETEC strains secreting the heat-stable toxin (ST) are the most
Arthur D Zimmerman et al.
Physiology & behavior, 227, 113150-113150 (2020-08-26)
Animals use social communication to learn important information from conspecifics that can guide appropriate behavioral choices. For example, during the social transmission of food preference (STFP), conspecific semiochemicals detected by mouse olfactory sensory neurons (OSNs) expressing the atypical olfactory receptor
Randy J Seeley et al.
The Journal of clinical investigation, 121(9), 3384-3386 (2011-08-26)
Prouroguanylin is a gastrointestinal paracrine signal and prohormone that is secreted after nutrient ingestion. In this issue of the JCI, Valentino et al. show that prouroguanylin is converted to uroguanylin in the CNS, which can activate guanylyl cyclase 2C (GUCY2C)
Matthew D Di Guglielmo et al.
Pediatric and developmental pathology : the official journal of the Society for Pediatric Pathology and the Paediatric Pathology Society, 21(3), 285-295 (2017-08-30)
Gastrointestinal tract-secreted satiety hormones play a significant role in one of the largest health-care challenges for children and adults, obesity. Recent studies in mice identified a novel role for uroguanylin, the endogenous intestinal hormone that binds guanylyl cyclase C (GUCY2C)
Martin Nue Møller et al.
The Laryngoscope, 127(6), E201-E208 (2017-03-16)
The function of the human endolymphatic sac (ES) has been enigmatic for decades. Hypotheses include controlling endolymphatic fluid homeostasis and inner ear immunological defense. Additionally, several studies indicate a possible endocrine capacity and a yet undefined role in intracranial pressure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service