Skip to Content
Merck
All Photos(1)

Key Documents

U3635

Sigma-Aldrich

Universal Antibody Dilution Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.47

pH

7.3-7.5

application(s)

hematology
histology

storage temp.

room temp

General description

Ready to use buffer for dilution of antibodies in all immunoassays. This buffer does not contain any mammalian proteins, phosphate, sodium azide or mercury preservatives and can be used for dilution of all antibodies, including peroxidase conjugates and antibodies to phosphoproteins. Not suitable for dilution of antibodies to S100 proteins.

Application

For dilution of primary or secondary antibodies. The diluted antibodies can be stored at 2-8 °C for several weeks in this buffer.
Universal Antibody Dilution Buffer has been used in immunoblotting.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Da Ma et al.
Oncology letters, 13(6), 4017-4026 (2017-06-08)
Calpain1, termed conventional calpain and a member of the Ca2+-dependent neutral cysteine proteases, is considered to be involved in cancer formation and development. However, the effect of calpain1 on oral squamous cell carcinoma (OSCC) remains poorly understood. The aim of
Zhihui Li et al.
Cell biology international, 46(10), 1672-1681 (2022-07-14)
Bladder cancer (BC) is a common urological malignancy that still lacks an effective treatment. Doxorubicin (Dox) has been widely used in the treatment of various cancers, including BC. However, chemoresistance often hampers the clinical application of Dox, therefore, it is
Caitlyn Fastenau et al.
Brain pathology (Zurich, Switzerland), 34(4), e13267-e13267 (2024-05-10)
Glycosylation is the most common form of post-translational modification in the brain. Aberrant glycosylation has been observed in cerebrospinal fluid and brain tissue of Alzheimer's disease (AD) cases, including dysregulation of terminal sialic acid (SA) modifications. While alterations in sialylation
Caitlyn Fastenau et al.
GeroScience, 45(3), 1539-1555 (2023-03-04)
Terminal sialic acid residues are present on most glycoproteins and glycolipids, but levels of sialylation are known to change in the brain throughout the lifespan as well as during disease. Sialic acids are important for numerous cellular processes including cell
High level of calpain1 promotes cancer cell invasion and migration in oral squamous cell carcinoma.
Ma D, et al.
Oncology Letters, 13(6), 4017-4026 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service