TRUESRGD
TrueGel3D Scramble RGD
Integrin Adhesion Peptide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
lyophilized powder
technique(s)
cell attachment: suitable
shipped in
dry ice
storage temp.
−20°C
General description
The scrambled RGD peptide has same amino acid content as RGD peptide but has low adhesion to integrin receptors and acts as control reagent to determine specificity of cell adhesion to RGD peptide. The thiol groups on the N-terminus acts as linker to bind non-adhesive scrambled RGD peptide to maleimide polymers (FAST-DEXTRAN/FAST-PVA).
Application
TRUESRGD is used as control reagent for RGD peptide to determine the specificity of cell adhesion caused by genuine RGD peptide (TrueGel3D RGD).
Features and Benefits
Contains a scrambled RGD sequence to serve as non-adhesive control peptide.
Components
TRUESRGD contains TrueGel3D scramble RGD peptide lyophilized and water.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service