Skip to Content
Merck
All Photos(1)

Key Documents

TRUEB

Sigma-Aldrich

TrueGel3D Buffer pH 7.2

600 μl solution, 10X concentrated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.21

shipped in

dry ice

storage temp.

−20°C

General description

TrueGel3D buffer pH 7.2 is a component of the TrueGel3D Hydrogel system. The TrueGel3D buffer with pH 7.2 is recommended for slow-gelling hydrogels, to ensure a homeostatic environment for cells. However, in conjugation with TrueGel3D buffer pH 5.5, buffer with intermediate strengths can be prepared to modulate gelation kinetics. Both buffer systems (TRUEA and TRUEB) have pH indicator (phenol red) and strong buffering capacity to ensure a stable pH.

Application

TrueGel3D buffers with pH 5.5 and 7.2 is used to modulate gelation kinetics

Components

TRUEB-1VL contains TrueGel3D buffer pH 7.2, 10X concentrated

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service